iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
Lieferant: AAT BIOQUEST
25561
AATB25561EA 144 EUR
AATB25561
iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
Proteinkonjugate und Markierungen
The iFluor® 700 conjugate of WGA possesses a bright, far-red fluorescence characteristic of the iFluor® 700 dye (Ex/Em = 690/713 nm).
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Bitte beachten For research use only.