Native Human Concanavalin A (from Human), iFluor® 488
Lieferant: AAT BIOQUEST
25580
AATB25580EA 143 EUR
AATB25580
Native Human Concanavalin A (from Human), iFluor® 488
Proteine und Peptide
Concanavalin A (ConA) is a lectin that binds specifically to certain structures found in various sugars, glycoproteins and glycolipids.
  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
Bitte beachten For research use only.