iFluor® 750-Concanavalin A Konjugat
Lieferant: AAT BIOQUEST
25598
AATB25598EA 144 EUR
AATB25598
iFluor® 750-Concanavalin A Konjugat
iFluor® 750-Concanavalin A
iFluor 750-labeled Concanavalin A selectively binds to a-mannopyranosyl and a-glucopyranosyl residues and exhibit the NIR fluorescence.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only