APC-iFluor™ 700 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert

Lieferant: AAT BIOQUEST
Achtung

2570
AATB2570EA 537 EUR
AATB2570
APC-iFluor™ 700 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
APC-iFluor™ 700 Tandem

Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.


  • Ready to use
  • High extinction coefficient
  • High quantum efficiency
  • Large Stokes shift

Molecular Weight: 105000 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR