APC-iFluor™ 750 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
Lieferant: AAT BIOQUEST
|
Achtung
|
2571
AATB2571EA
537
EUR
AATB2571
APC-iFluor™ 750 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
APC-iFluor™ 750 Tandem
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
- Ready to use
- High extinction coefficient
- High quantum efficiency
- Large Stokes shift
- Spirulina
Molecular Weight:
105000 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...