APC-iFluor™ 800 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
Lieferant: AAT BIOQUEST
Achtung
2572
AATB2572EA 527 EUR
AATB2572
APC-iFluor™ 800 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
APC-iFluor™ 800 Tandem
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
  • Ready to use
  • High extinction coefficient
  • High quantum efficiency
  • Large Stokes shift
ADR: 5,II