PE-iFluor™ 647 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
Lieferant: AAT BIOQUEST
|
Achtung
|
2577
AATB2577EA
537
EUR
AATB2577
PE-iFluor™ 647 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
PE-iFluor™ 647 Tandem
R-Phycoerythrin (PE) is isolated from red algae.
- Ready to use
- Excellent conjugation yield
Molecular Weight:
240000 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...