PE-iFluor™ 750 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert

Lieferant: AAT BIOQUEST
Achtung

2578
AATB2578EA 537 EUR
AATB2578
PE-iFluor™ 750 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
PE-iFluor™ 750 Tandem

R-Phycoerythrin (PE) is isolated from red algae.


  • Ready to use
  • Excellent conjugation yield

Molecular Weight: 240000 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR