PE-iFluor™ 660 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
Lieferant: AAT BIOQUEST
|
Achtung
|
2579
AATB2579EA
527
EUR
AATB2579
PE-iFluor™ 660 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
PE-iFluor™ 660 Tandem
R-Phycoerythrin (PE) is isolated from red algae.
- Ready to use
- Excellent conjugation yield
Molecular Weight:
1960000 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...