PE-iFluor™ 594 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert

Lieferant: AAT BIOQUEST
Achtung

2584
AATB2584EA 537 EUR
AATB2584
PE-iFluor™ 594 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
PE-iFluor™ 594 Tandem

PE-Texas Red is a popular color used in flow cytometry.


  • Ready to use
  • Excellent conjugation yield

Molecular Weight: 240000 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR