APC-Cy5.5 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert

Lieferant: AAT BIOQUEST
Achtung

2586
AATB2586EA 537 EUR
AATB2586
APC-Cy5.5 Tandem, ReadiUse™ fluoreszierende Probe, voraktiviert
APC-Cy5.5 Tandem

Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.


  • Ready to use
  • Excellent conjugation yield

Molecular Weight: 105000 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR