PE-iFluor® 594 Tandem
Lieferant: AAT BIOQUEST
2600
AATB2600EA 304 EUR
AATB2600
PE-iFluor® 594 Tandem
PE-iFluor® 594
PE-iFluor® 594 is a superior replacement to the commonly used PE-Texas Red tandem with significantly improved FRET efficiency and signal/background ratio.
  • Stable for 6 months
Tandem dyes are a unique class of fluorescent molecules that consist of two different covalently linked fluorophores.
Bitte beachten For Research Use Only