N-Me-DMAE NHS ester
Lieferant: AAT Bioquest
26005
AATB26005EA 307 EUR
AATB26005
N-Me-DMAE NHS ester
N-Me-DMAE NHS ester
The acridinium NHS ester can be used to label proteins and nucleic acids.
  • Chemiluminescent labeling
  • Excellent sensitivity
  • Suitable for various Experimental surfaces
  • Multiple applications
Bitte beachten For research use only.