N-Me-DMAE NHS ester
Lieferant: AAT Bioquest
26005
AATB26005EA
307
EUR
AATB26005
N-Me-DMAE NHS ester
N-Me-DMAE NHS ester
The acridinium NHS ester can be used to label proteins and nucleic acids.
- Chemiluminescent labeling
- Excellent sensitivity
- Suitable for various Experimental surfaces
- Multiple applications
Bitte beachten For research use only.