PE-iFluor® 597 Tandem
Lieferant: AAT BIOQUEST
2601
AATB2601EA
304
EUR
AATB2601
PE-iFluor® 597 Tandem
PE-iFluor® 597
PE-iFluor® 597 is a superior replacement to the commonly used PE-Texas Red tandem with significantly improved FRET efficiency and signal/background ratio.
- Stable for 6 months
Tandem dyes are a unique class of fluorescent molecules that consist of two different covalently linked fluorophores.
Bitte beachten For Research Use Only