PE-iFluor® 660 Tandem
Lieferant: AAT BIOQUEST
2602
AATB2602EA 304 EUR
AATB2602
PE-iFluor® 660 Tandem
PE-iFluor® 660
PE-iFluor® 660 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
  • Stable for 6 months
Tandem dyes are a unique class of fluorescent molecules that consist of two different covalently linked fluorophores.
Bitte beachten For Research Use Only