APC-Cy5.5 Tandem fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

2622
AATB2622EA 307 EUR
AATB2622
APC-Cy5.5 Tandem fluoreszierende Probe
APC-Cy5.5 Tandem

Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green algae.


  • Suitable for most fluorescence platforms
  • Excellent signal
  • High quality

Molecular Weight: 105000 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR