APC-iFluor® 700 Tandem
Lieferant: AAT BIOQUEST
2623
AATB2623EA 304 EUR
AATB2623
APC-iFluor® 700 Tandem
APC-iFluor® 700
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
  • Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only