APC-XFD700 Tandem
Lieferant: AAT BIOQUEST
2624
AATB2624EA
304
EUR
AATB2624
APC-XFD700 Tandem
APC-XFD700
Allophycocyanin (APC) is a phycobiliprotein with an extremely high absorptivity and a high quantum efficiency.
- Stable for 6 months
APC-XFD 700 gives significantly improved stain index for certain CD antibodies than APC-Alexa Fluor® 700 tandem under the same conditions.
Bitte beachten For Research Use Only