APC-iFluor® 750 Tandem
Lieferant: AAT BIOQUEST
2626
AATB2626EA
304
EUR
AATB2626
APC-iFluor® 750 Tandem
APC-iFluor® 750
APC-iFluor® 750 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only