APC-iFluor® 800 Tandem
Lieferant: AAT BIOQUEST
2630
AATB2630EA
304
EUR
AATB2630
APC-iFluor® 800 Tandem
APC-iFluor® 800
APC-iFluor® 800 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only