APC-iFluor®A7 Tandem
Lieferant: AAT BIOQUEST
2632
AATB2632EA
348
EUR
AATB2632
APC-iFluor®A7 Tandem
APC-iFluor®A7
APC-iFluor® A7 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only