APC-iFluor® 740 Tandem
Lieferant: AAT BIOQUEST
2635
AATB2635EA 304 EUR
AATB2635
APC-iFluor® 740 Tandem
APC-iFluor® 740
APC-iFluor® 740 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
  • Stable for 6 months
Quality tested in-house