APC-iFluor® 770 Tandem
Lieferant: AAT BIOQUEST
2638
AATB2638EA
304
EUR
AATB2638
APC-iFluor® 770 Tandem
APC-iFluor® 770
APC-iFluor® 770 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only