APC-iFluor® 780 Tandem
Lieferant: AAT BIOQUEST
2639
AATB2639EA
304
EUR
AATB2639
APC-iFluor® 780 Tandem
APC-iFluor® 780
APC-iFluor® 780 is a new unique tandem color for multicolor applications, in particular, for designing spectral flow cytometric panels.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only