Cyanine 3 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy3® bisNHS ester]

Lieferant: AAT BIOQUEST

270
AATB270EA 307 EUR
AATB270
Cyanine 3 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy3® bisNHS ester]
Cyanine 3 bissuccinimidyl ester potassium salt

This Cy3® dye is the same molecule to GE's Cy3® bisNHS ester.


  • Multiple Bioconjugation Applications
  • 90% Purity


Bitte beachten For research use only.

Formel: C43H49KN4O14S2
CAS-Nummer: 1311966-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR