PE-iFluor® 647 Tandem
Lieferant: AAT BIOQUEST
2702
AATB2702EA
304
EUR
AATB2702
PE-iFluor® 647 Tandem
PE-iFluor® 647
PE-iFluor® 647 is a superior replacement to the commonly used PE-Cy5 tandem with significantly improved FRET efficiency and signal/background ratio.
- Stable for 6 months
Tandem dyes are a unique class of fluorescent molecules that consist of two different covalently linked fluorophores.
Bitte beachten For Research Use Only