Cyanine 3 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3® NHS ester]
Lieferant: AAT BIOQUEST
271
AATB271EA 152 EUR
AATB271
Cyanine 3 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3® NHS ester]
Cyanine 3 monosuccinimidyl ester potassium salt
This Cy3® dye is the same molecule to GE's Cy3® NHS ester.
  • Multiple Bioconjugation Applications
  • 90% Purity
Bitte beachten For research use only.
CAS-Nummer: 146368-16-3