PE-Cy5.5 maleimide, ReadiUse™, voraktiviert
Lieferant: AAT BIOQUEST
2718
AATB2718EA
561
EUR
AATB2718
PE-Cy5.5 maleimide, ReadiUse™, voraktiviert
PE-Cy5.5 maleimide
PE-Cy5.5 is a popular tandem color used in flow cytometry.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only