PerCP maleimide, ReadiUse™, voraktiviert
Lieferant: AAT BIOQUEST
2723
AATB2723EA 561 EUR
AATB2723
PerCP maleimide, ReadiUse™, voraktiviert
PerCP maleimide
PerCP (Peridinin-chlorophyll-protein complex) is isolated from Dinophyceae sp.
  • Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only