Cyanine 3.5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3.5® NHS ester]
Lieferant: AAT BIOQUEST
275
AATB275EA
307
EUR
AATB275
Cyanine 3.5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3.5® NHS ester]
Cyanine 3.5 monosuccinimidyl ester potassium salt
This Cy3.5® dye is the same molecule to GE's Cy3.5® NHS ester.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.