Cyanine 5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy5® NHS ester]
Lieferant: AAT BIOQUEST
280
AATB280EA
152
EUR
AATB280
Cyanine 5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy5® NHS ester]
Cyanine 5 monosuccinimidyl ester potassium salt
This Cy5® dye is the same molecule to GE's monoreactive Cy5® NHS ester.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.
CAS-Nummer:
146368-14-1 |