ATTO 488 acid
Lieferant: AAT BIOQUEST
2813
AATB2813EA
464
EUR
AATB2813
ATTO 488 acid
ATTO 488 acid
ATTO 488 is a hydrophilic, rhodamine-based fluorescent dye with exceptional water solubility.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only