ATTO 488 acid
Lieferant: AAT BIOQUEST
2813
AATB2813EA 464 EUR
AATB2813
ATTO 488 acid
ATTO 488 acid
ATTO 488 is a hydrophilic, rhodamine-based fluorescent dye with exceptional water solubility.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only