ATTO 488 NHS ester
Lieferant: AAT BIOQUEST
2815
AATB2815EA
153
EUR
AATB2815
ATTO 488 NHS ester
ATTO 488 NHS ester
ATTO 488 is a hydrophilic, rhodamine-based fluorescent dye with exceptional water solubility.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only