ATTO 532 acid
Lieferant: AAT BIOQUEST
2820
AATB2820EA
464
EUR
AATB2820
ATTO 532 acid
ATTO 532 acid
ATTO 532 is a rhodamine-based fluorescent labeling dye with high molecular high extinction coefficient and fluorescence quantum yield (0.90) as well as sufficient Stoke's shift (excitation maximum 532 nm, emission maximum 553 nm).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only