ATTO 532 NHS ester
Lieferant: AAT BIOQUEST
2822
AATB2822EA
153
EUR
AATB2822
ATTO 532 NHS ester
ATTO 532 NHS ester
ATTO 532 is a rhodamine-based fluorescent labeling dye with high molecular high extinction coefficient and fluorescence quantum yield (0.90) as well as sufficient Stoke's shift (excitation maximum 532 nm, emission maximum 552 nm).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only