ATTO 565 maleimide
Lieferant: AAT BIOQUEST
2829
AATB2829EA 130 EUR
AATB2829
ATTO 565 maleimide
ATTO 565 maleimide
ATTO 565 Maleimide is a commonly used thiol-reactive rhodamine dye.
  • Stable for 24 months
Quality tested in-house