Cyanine 5.5 monosuccinimidyl ester potassium salt ≥90% [same as Cy5.5® NHS ester]
Lieferant: AAT BIOQUEST
283
AATB283EA 307 EUR
AATB283
Cyanine 5.5 monosuccinimidyl ester potassium salt ≥90% [same as Cy5.5® NHS ester]
Cyanine 5.5 monosuccinimidyl ester potassium salt
This Cy5.5® dye is the same molecule to GE's monoreactive Cy5.5® NHS ester.
  • Multiple Bioconjugation Applications
  • 90% Purity
Bitte beachten For research use only.