ATTO 647 acid
Lieferant: AAT BIOQUEST
2850
AATB2850EA
775
EUR
AATB2850
ATTO 647 acid
ATTO 647 acid
ATTO 647 is a rhodamine-derived fluorescent dye optimized for applications within the red spectral region, offering spectral characteristics similar to Cy5.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only