ATTO 647 NHS ester
Lieferant: AAT BIOQUEST
2852
AATB2852EA
309
EUR
AATB2852
ATTO 647 NHS ester
ATTO 647 NHS ester
ATTO 647 is a rhodamine-derived fluorescent dye optimized for applications within the red spectral region, offering spectral characteristics similar to Cy5.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only