ATTO 647N NHS ester
Lieferant: AAT BIOQUEST
2856
AATB2856EA
309
EUR
AATB2856
ATTO 647N NHS ester
ATTO 647N NHS ester
ATTO 647N is a rhodamine-based fluorescent dye optimized for use in the red spectral region, with similar spectral characteristics as Cy5.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only