Atto 647N maleimide
Lieferant: AAT BIOQUEST
2857
AATB2857EA
309
EUR
AATB2857
Atto 647N maleimide
Atto 647N maleimide
ATTO 647N is a rhodamine-based fluorescent dye optimized for use in the red spectral region, with similar spectral characteristics as Cy5.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only