Atto 594 maleimide
Lieferant: AAT BIOQUEST
2861
AATB2861EA 309 EUR
AATB2861
Atto 594 maleimide
Atto 594 maleimide
ATTO 594 is a rhodamine-based fluorescent label known for its excellent water solubility.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only