Atto 594 maleimide
Lieferant: AAT BIOQUEST
2861
AATB2861EA
309
EUR
AATB2861
Atto 594 maleimide
Atto 594 maleimide
ATTO 594 is a rhodamine-based fluorescent label known for its excellent water solubility.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only