ATTO 514 acid
Lieferant: AAT BIOQUEST
2862
AATB2862EA 464 EUR
AATB2862
ATTO 514 acid
ATTO 514 acid
ATTO 514 is a rhodamine-based fluorophore characterized by exceptional water solubility, strong absorption, high fluorescence quantum yield, and outstanding thermal and photo-stability.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only