ATTO 514 NHS ester
Lieferant: AAT BIOQUEST
2863
AATB2863EA 153 EUR
AATB2863
ATTO 514 NHS ester
ATTO 514 NHS ester
ATTO 514, a hydrophilic rhodamine-based fluorophore, is known for its excellent water solubility, strong absorption, high fluorescence quantum yield, and thermal and photo-stability.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only