Cyanine 7 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy7® NHS ester]

Lieferant: AAT BIOQUEST

Synonyme: Cy7 NHS ester

290
AATB290EA 307 EUR
AATB290
Cyanine 7 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy7® NHS ester]
Cyanine 7 monosuccinimidyl ester potassium salt

This Cy7® dye is the same molecule to GE's monoreactive Cy7® NHS ester.


  • Multiple Bioconjugation Applications
  • 90% Purity


Bitte beachten For research use only.

CAS-Nummer: 477908-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR