D(+)-Biotin ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: (3aS,4S,6aR)-Biotin, Vitamin H, Vitamin B8, 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H- thieno[3,4-d]imidazol-4-yl]pentansäure, Vitamin B7, Coenzym R

3001
AATB3001EA 120 EUR
AATB3001
D(+)-Biotin ≥90%
D(+)-Biotin

Biotin binds with high affinity to both avidins and streptavidins.

Formel: C₁₀H₁₆N₂O₃S
Molecular Weight: 244,31 g/mol
MDL: MFCD00005541
CAS-Nummer: 58-85-5
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR