1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

3002
AATB3002EA 307 EUR
AATB3002
1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion ≥90%
D-Biotin-N-succinimidyl ester

Biotin succinimidyl ester is the most popular amine-reactive biotin derivative for modifying proteins and other biological molecules.

Formel: C₁₄H₁₉N₃O₅S
Molecular Weight: 341,38 g/mol
MDL: MFCD00078531
CAS-Nummer: 35013-72-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR