N-Biotinyl-ethylenediamine trifluoroacetate salt für die Molekularbiologie

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-(2-Aminoethyl)biotinamide trifluoroacetate salt

3003
AATB3003EA 153 EUR
AATB3003
N-Biotinyl-ethylenediamine trifluoroacetate salt für die Molekularbiologie
N-Biotinyl-ethylenediamine trifluoroacetate salt

Biotin ethylenediamine is widely used to label biomolecules that contain either carboxy, phosphonyl or carbonyl group.


  • Useful for multiple applications

Formel: C₁₂H₂₂N₄O₂S·C₂HF₃O₂
Molecular Weight: 400,42 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11045268
CAS-Nummer: 1217450-40-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR