Biotin cadaverine kolorimetrische Tests

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-(5-Aminopentyl)biotinamide, trifluoroacetic acid salt

3004
AATB3004EA 152 EUR
AATB3004
Biotin cadaverine kolorimetrische Tests
Biotin cadaverine

Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.


  • Carbonyl-reactive
  • Multiple applications

Formel: C₁₇H₂₉F₃N₄O₄S
Molecular Weight: 442,5 g/mol
MDL: MFCD03452814
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a9b7984de22acf84dfaa1b7c1ed1a89e82feb90&vid=8e61c368b6758f050777ebf07b943bf3f16c356a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR