Biotin C2 maleimide Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Biotin-C2-maleimide, N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide
3005
AATB3005EA
460
EUR
AATB3005
Biotin C2 maleimide Biodetektion
Biotin C2 maleimide
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.
- Thiol-reactive
- Multiple applications
Formel:
C₁₆H₂₂SN₄O₄ Molecular Weight: 366,3 g/mol |
MDL:
MFCD02094223 CAS-Nummer: 139554-72-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...